930059
S-Phenyl methanethioate
Synonym(s):
Phenyl thioCORMate, Phenyl thioformate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
Quality Level
refractive index
n/D 1.586
density
1.134 g/mL
functional group
thioether
storage temp.
−20°C
SMILES string
[H]C(SC1=CC=CC=C1)=O
InChI
1S/C7H6OS/c8-6-9-7-4-2-1-3-5-7/h1-6H
InChI key
ITJOOMDVHPTIRB-UHFFFAOYSA-N
Application
S-phenyl methanethioate is an effective CO releasing molecule (CORM) developed by the lab of Sarah Tasker as part of a series of thioformate based CORMs called thioCORMates. p-NO2 phenyl thioformate generates CO over the course of several minutes to allow for on-demand generation of CO for variety of reactions.
This and the other thioCORMates are cost effective CORMs that when activated with simple amines, can be used to generate a near quantitative amount of CO safely without the usage of gas cylinders. These are compatible with a variety of carbonylation reactions, as well as existing COware glassware and components.
ThioCORMate: Tunable and Cost-Effective Carbon Monoxide-Releasing Molecule
Learn more about CO releasing molecules from Prof. Skrydstrup
This and the other thioCORMates are cost effective CORMs that when activated with simple amines, can be used to generate a near quantitative amount of CO safely without the usage of gas cylinders. These are compatible with a variety of carbonylation reactions, as well as existing COware glassware and components.
ThioCORMate: Tunable and Cost-Effective Carbon Monoxide-Releasing Molecule
Learn more about CO releasing molecules from Prof. Skrydstrup
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
221.0 °F
Flash Point(C)
105 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service