Skip to Content
Merck
All Photos(2)

Key Documents

905240

Sigma-Aldrich

(R,R,R)-SPIRAP

≥95%

Synonym(s):

((1R,3R,3′R)-3,3′-diethyl-3H,3′H-1,1′-spirobi[isobenzofuran]-7,7′-diyl)bis(diphenylphosphane), (R)-CrabPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H38O2P2
Molecular Weight:
648.71
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: ligand
reaction type: Asymmetric synthesis

mp

226-228 °C

SMILES string

CC[C@@H](O1)C2=C([C@@]1(O[C@@H]3CC)C4=C3C=CC=C4P(C5=CC=CC=C5)C6=CC=CC=C6)C(P(C7=CC=CC=C7)C8=CC=CC=C8)=CC=C2

InChI

1S/C43H38O2P2/c1-3-37-35-27-17-29-39(46(31-19-9-5-10-20-31)32-21-11-6-12-22-32)41(35)43(44-37)42-36(38(4-2)45-43)28-18-30-40(42)47(33-23-13-7-14-24-33)34-25-15-8-16-26-34/h5-30,37-38H,3-4H2,1-2H3/t37-,38-,43-/m1/s1

InChI key

PRORVWYPBRMLLA-FFUXEPPCSA-N

Application

(R,R,R)-SPIRAP is a chiral C2-symmetric spiroketal-containing ligand developed in the Nagorny lab for asymmetric catalysis. It can be used in variety of stereoselective transformations, including: Ir-catalyzed hydroarylation, Pd-catalyzed allylic alkylation, Pd-catalyzed Heck couplings, and Rh-catalyzed hydrogenation.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alonso J Argüelles et al.
Angewandte Chemie (International ed. in English), 57(19), 5325-5329 (2018-02-24)
We present an expedient and economical route to a new spiroketal-based C2 -symmetric chiral scaffold, termed SPIROL. Based on this spirocyclic scaffold, several chiral ligands were generated. These ligands were successfully employed in an array of stereoselective transformations, including in
Design, Synthesis, and Application of Chiral C2-Symmetric Spiroketal-Containing Ligands in Transition-Metal Catalysis.
Arguelles AJ, et al.
Angewandte Chemie (International Edition in English), 57(19), 5325-5329 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service