Skip to Content
Merck
All Photos(1)

Key Documents

924024

Sigma-Aldrich

Silver bromide

≥99.9% trace metals basis

Synonym(s):

Argentous bromide, Silver monobromide, Silver(I) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgBr
CAS Number:
Molecular Weight:
187.77
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Assay:
≥99.9% trace metals basis
form:
powder, crystals or chunks

Quality Level

Assay

≥99.9% trace metals basis

form

powder, crystals or chunks

mp

432 °C (lit.)

density

6.473 g/mL at 25 °C (lit.)

SMILES string

Br[Ag]

InChI

1S/Ag.BrH/h;1H/q+1;/p-1

InChI key

ADZWSOLPGZMUMY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Silver bromide (AgBr) with 99.9% trace metal purity is an inorganic compound employed in diverse fields due to its remarkable light sensitivity, optical properties, and high purity.

Application

Silver bromide (AgBr) is a promising photocatalyst with a narrow bandgap. Three-dimensional hydrogels incorporating AgBr exhibit exceptional antibacterial properties, making them ideal for photochromic contact lenses. Graphene-based aerogels used as catalyst supports for AgBr have shown high photocatalytic activity, effective in the photodegradation of organic pollutants and antibacterial applications. Additionally, AgBr can be used to synthesize silver bromide nanoparticles, which have applications in catalysis, sensing, and photonic devices. Silver bromide is also utilized in research for creating thin films and coatings with unique optical and electronic properties.

Features and Benefits

Silver bromide with 99.9% trace metal purity minimizes contamination from other metals, making it perfect for high-precision and high-performance applications. This exceptional purity ensures consistent and reliable properties, which are essential for scientific research and advanced industrial processes.

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Review on advances in photocatalytic water disinfection utilizing graphene and graphene derivatives-based nanocomposites
Dutta V, et al.
Journal of Environmental Chemical Engineering, 7(3), 103132-103132 (2019)
Construction of Z-scheme and p-n heterostructure: Three-dimensional porous g-C3N4/graphene oxide-Ag/AgBr composite for high-efficient hydrogen evolution
Li W, et al.
Applied Catalysis. B, Environmental, 268, 118384-118384 (2020)
Hydrogen evolution by catalyzing water splitting on two-dimensional g-C3N4-Ag/AgBr heterostructure
Wei L, et al.
Applied Surface Science, 494, 275-284 (2019)
RETRACTED: Z-scheme g-C3N4nanosheet/MgBi2O6 systems with the visible light response for impressive photocatalytic organic contaminants degradation
Nguyen V H, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 406, 113023-113023 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service