Skip to Content
Merck
All Photos(4)

Documents

204382

Sigma-Aldrich

Silver chloride

99.999% trace metals basis

Synonym(s):

Chlorosilver

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgCl
CAS Number:
Molecular Weight:
143.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 912 °C)

Assay

99.999% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: silver

impurities

≤15.0 ppm Trace Metal Analysis

refractive index

n20/D 2.0668 (lit.)

mp

455 °C (lit.)

SMILES string

Cl[Ag]

InChI

1S/Ag.ClH/h;1H/q+1;/p-1

InChI key

HKZLPVFGJNLROG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Applied in controlled assembly of one-dimensional and three-dimensional silver(I) thiolates.
Used with palladium in a catalytic cross-coupling of silylacetylenes with aryl halides.

Pictograms

CorrosionEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlled Assembly Based on Multibridging Thiolate Ligands: New Polymeric Silver(I) Complexes with One-Dimensional Chain and Three-Dimensional Network Structures.
Maochun Hong et al.
Inorganic chemistry, 38(3), 600-602 (2001-10-25)
Synthetic Communications, 37, 1301-1301 (2007)
Edward M Kosower et al.
Langmuir : the ACS journal of surfaces and colloids, 28(37), 13208-13217 (2012-08-31)
Evaporation of water on a planar AgX surface leads to a strongly bound monolayer for which IR spectra display the marker peaks for modest numbers of oligomers. From 700-1800 spectra for each isotopomer, H(2)O(16) and H(2)O(18), a pair was selected
Qiong Wang et al.
Biosensors & bioelectronics, 43, 19-24 (2013-01-01)
In this work, we describe a new sensitive strategy for electrochemical detection of protein via small molecule/protein interactions. This assay is based on a terminal protection mechanism that small molecule-linked single-stranded DNA (ssDNA) is protected against hydrolysis by exonuclease I
Siti Fauziyah Rahman et al.
Journal of microbiology and biotechnology, 22(10), 1446-1451 (2012-10-19)
Levodopa or L-3,4-dihydroxyphenylalanine (L-DOPA) is the precursor of the neurotransmitter dopamine. L-DOPA is a famous treatment for Parkinson's disease symptoms. In this study, electroenzymatic synthesis of L-DOPA was performed in a three-electrode cell, comprising a Ag/AgCl reference electrode, a platinum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service