Skip to Content
Merck
All Photos(1)

Key Documents

918067

Sigma-Aldrich

Indium tin oxide coated PEN with titanium(IV) oxide buffer layer

>80% transparency, film L × W × thickness 29.7 cm × 21 cm × 125 μm, sheet resistance <15 Ω/sq

Synonym(s):

ITO coated polyethylene naphthalate with titanium dioxide buffer layer, ITO-PEN with TiO2 buffer layer (>80% transparency)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

material

substrate (Polyethylene naphlalate (PEN) film (one side coated) )

Quality Level

sheet resistance

<15 Ω/sq

film L × W × thickness

29.7 cm × 21 cm × 125 μm

layer thickness

7 nm , TiO2

transmittance

550 nm, >80%

Application

This product is an one-side coated, flexible, ITO coated PEN polymer film. It is a highly-transparent, ready-to-use, flexible conductive substrate with low sheet resistance. Titanium dioxide buffer layer was coated onto the coated side of the film with >80% transmittance. This product can be used for rapid fabrication of flexible Perovskite solar cells and flexible dye-sensitized solar cells.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photovoltaic Performance of Plastic Dye-Sensitized Electrodes Prepared by Low-Temperature Binder-Free Coating of Mesoscopic Titania
Miyasaka T et al
Journal of the Electrochemical Society, 154 (2007)
Jennifer M Pringle et al.
Chemical communications (Cambridge, England), 46(29), 5367-5369 (2010-06-23)
Electrodeposition of poly(3,4-ethylenedioxythiophene) onto conducting plastic allows the cheap and facile synthesis of plastic cathodes for dye-sensitised solar cells, having excellent solar cell efficiency (8%), at a fraction of the cost of platinised plastic.
Low temperature fabrication of high performance and transparent Pt counter electrodes for use in flexible dye-sensitized solar cells.
Xiao Y, et al.
Science Bulletin, 57, 2329-2334 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service