Skip to Content
Merck
All Photos(1)

Documents

915181

Sigma-Aldrich

Cesium bromide

AnhydroBeads, 99.999% trace metals basis, (Perovskite grade)

Synonym(s):

Cesium monobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsBr
CAS Number:
Molecular Weight:
212.81
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

form

solid

particle size

-10 mesh

mp

636 °C (lit.)

density

4.44 g/mL at 25 °C (lit.)

SMILES string

[Br-].[Cs+]

InChI

1S/BrH.Cs/h1H;/q;+1/p-1

InChI key

LYQFWZFBNBDLEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cesium bromide (CsBr) is commonly used in scintillation detectors, which are devices that detect and measure ionizing radiation. CsBr has also found applications in optoelectronic devices.
Cesium bromide finds application in synthesis of perovksites based photovoltaic materials. Our perovskite grade CsBr can readily be dissolved in DMF to yield 1M solution.

Packaging

Packaged in ampules

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dandan Zhang et al.
Journal of the American Chemical Society, 137(29), 9230-9233 (2015-07-17)
Halide perovskites have attracted much attention over the past 5 years as a promising class of materials for optoelectronic applications. However, compared to hybrid organic-inorganic perovskites, the study of their pure inorganic counterparts, like cesium lead halides (CsPbX3), lags far
Michael Kulbak et al.
The journal of physical chemistry letters, 7(1), 167-172 (2015-12-25)
Direct comparison between perovskite-structured hybrid organic-inorganic methylammonium lead bromide (MAPbBr3) and all-inorganic cesium lead bromide (CsPbBr3), allows identifying possible fundamental differences in their structural, thermal and electronic characteristics. Both materials possess a similar direct optical band gap, but CsPbBr3 demonstrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service