912883
tBuBrettPhos Pd G6 TES
Synonym(s):
tBuBrettPhos OAC precatalyst TES
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
form
powder
Quality Level
reaction suitability
reagent type: catalyst
reaction type: Cross Couplings
mp
>300 °C
SMILES string
[Pd](Br)c3ccc(cc3)C(=O)OCC[Si](C)(C)C.P(C(C)(C)C)(C(C)(C)C)c1c(ccc(c1c2c(cc(cc2C(C)C)C(C)C)C(C)C)OC)OC
InChI key
ZIQMRJCMTGYLPQ-UHFFFAOYSA-M
Application
tBuBrettPhos Pd G6 TES is a G6 Buchwald precatalyst. The G6 precatalysts are air-stable, palladium-based oxidative addition complexes (OACs) that offer an alternative to the previously developed classes of palladacycle precatalysts. Unlike previous generations, the bulkiest biarylphosphine ligand precatalysts are readily prepared. These precatalysts have been shown to be effective for many reactions, including C-N, C-O, and C-F cross couplings.
Learn more about G6 Buchwald precatalysts
Learn more about G6 Buchwald precatalysts
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Monophosphine Ligands Promote Pd-Catalyzed C-S Cross-Coupling Reactions at Room Temperature with Soluble Bases.
ACS Catalysis, 9(7), 6461-6466 (2019)
Organic letters, 19(11), 2853-2856 (2017-05-13)
In this report, we describe the application of palladium-based oxidative addition complexes (OACs) as effective precatalysts for C-N, C-O, and C-F cross-coupling reactions with a variety of (hetero)arenes. These complexes offer a convenient alternative to previously developed classes of precatalysts
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service