Skip to Content
Merck
All Photos(5)

Key Documents

745979

Sigma-Aldrich

tBuBrettPhos Pd G3

96%

Synonym(s):

tert-BuBrettPhos-Pd-G3, [(2-Di-tert-butylphosphino-3,6-dimethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H62NO5PPdS
CAS Number:
Molecular Weight:
854.43
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

119-131 °C

functional group

phosphine

SMILES string

COC1=CC=C(C(P(C(C)(C)C)C(C)(C)C)=C1C2=C(C=C(C=C2C(C)C)C(C)C)C(C)C)OC.NC3=C(C=CC=C3)C4=C(C=CC=C4)[Pd]OS(C)(=O)=O

InChI

1S/C31H49O2P.C12H10N.CH4O3S.Pd/c1-19(2)22-17-23(20(3)4)27(24(18-22)21(5)6)28-25(32-13)15-16-26(33-14)29(28)34(30(7,8)9)31(10,11)12;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h15-21H,1-14H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

GAQPAUHHECNHNS-UHFFFAOYSA-M

General description

tBuBrettPhos Pd G3 is a third generation (G3) Buchwald precatalyst that can be used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. It is air-, moisture-, and thermally-stable and is highly soluble in a wide range of common organic solvents. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

Application

tBuBrettPhos Pd G3 has been used as a precatalyst for the N-arylation of amino acid esters with aryl triflates under mild reaction conditions and minimal racemization of the amino acid ester. It may also be used to catalyze the conversion of aryl halides to phenols in the presence of benzaldoxime as a hydroxide surrogate.

For small scale and high throughput uses, product is also available as ChemBeads (928313)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hailey A Young et al.
The Journal of organic chemistry, 85(3), 1748-1755 (2019-12-04)
Palladium-catalyzed N-arylations of amino acid tert-butyl esters using 4-bromo-N,N-dimethylaniline as a coupling partner are reported. The resulting N-aryl amino acid esters are suitable building blocks for the synthesis of electron-rich N-aryl peptides, which undergo oxidative couplings to aminooxy groups to
Synthesis of Complex Phenols Enabled by a Rationally Designed Hydroxide Surrogate.
Fier PS & Maloney KM
Angewandte Chemie (International Edition in English), 56(16), 4478-4482 (2017)
Sandra M King et al.
Organic letters, 18(16), 4128-4131 (2016-08-09)
A general method for the N-arylation of amino acid esters with aryl triflates is described. Both α- and β-amino acid esters, including methyl, tert-butyl, and benzyl esters, are viable substrates. Reaction optimization was carried out by design of experiment (DOE)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service