All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H19NO5
CAS Number:
Molecular Weight:
209.24
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥98.0%
form
powder
optical activity
[α]/D −17.0±2.0°, c = 1% in H2O
mp
136-142 °C
solubility
H2O: 0.1 g/mL, clear to almost clear, colorless to very faintly yellow
SMILES string
CCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
InChI
1S/C8H19NO5/c1-2-9-3-5(11)7(13)8(14)6(12)4-10/h5-14H,2-4H2,1H3/t5-,6+,7+,8+/m0/s1
InChI key
IKXCHOUDIPZROZ-LXGUWJNJSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service