H8896
cis-10-Heptadecenoic acid
≥99%
Synonym(s):
(10Z)-10-Heptadecenoic acid, 10-cis-Heptadecenoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H32O2
CAS Number:
Molecular Weight:
268.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99%
storage temp.
−20°C
SMILES string
CCCCCC\C=C/CCCCCCCCC(O)=O
InChI
1S/C17H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h7-8H,2-6,9-16H2,1H3,(H,18,19)/b8-7-
InChI key
GDTXICBNEOEPAZ-FPLPWBNLSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sarah M Grundmann et al.
Scientific reports, 10(1), 20748-20748 (2020-11-29)
The consumption of phosphorus in Western populations largely exceeds the recommended intake, while vitamin D supply is often insufficient. Both situations are linked to an increased cardiovascular risk. A 17-week two-factorial study with Ldl receptor-/- mice was conducted to investigate the
Renato B Pereira et al.
Chemosphere, 208, 682-690 (2018-06-13)
The profile of essential and non-essential elements was traced in the edible sea hares Aplysia depilans Gmelin, Aplysia fasciata Poiret and Aplysia punctata Cuvier. Manganese (Mn), iron (Fe), zinc (Zn), copper (Cu) and selenium (Se) were identified as the major
Guangling Jiao et al.
Marine drugs, 13(6), 3849-3876 (2015-06-23)
Northern shrimp (Pandalus borealis) oil, which is rich in omega-3 fatty acids, was recovered from the cooking water of shrimp processing facilities. The oil contains significant amounts of omega-3 fatty acids in triglyceride form, along with substantial long-chain monounsaturated fatty
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service