Skip to Content
Merck
All Photos(2)

Key Documents

906336

Sigma-Aldrich

PBDB-T-2F

Synonym(s):

PCE135, PM6, Poly[[4,8-bis[5-(2-ethylhexyl)-4-fluoro-2-thienyl]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl]-2,5-thiophenediyl[5,7-bis(2-ethylhexyl)-4,8-dioxo-4H,8H-benzo[1,2-c:4,5-c′]dithiophene-1,3-diyl]-2,5-thiophenediyl]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C68H76F2O2S8)n
CAS Number:
UNSPSC Code:
12352101
NACRES:
NA.23

description

Band gap: 1.9 eV

form

particles

mol wt

Mw 80,000-200,000 g/mol by GPC

color

Fine

solubility

chlorobenzene: soluble
chloroform: soluble
dichlorobenzene: soluble

Orbital energy

HOMO -5.5 eV 
LUMO -3.6 eV 

PDI

2‑4

Application

High-Efficiency Organic Solar Cells (OPVs)
Polymeric donor material
LUMO=−3.6 eV
HOMO=−5.5 eV

OPV Device Performance:

PBDB-T-2F: ITIC-F (1:1 w/w)
Voc= 0.84V
Jsc= 22.2 mA/cm2
FF= 0.725
PCE=13.5%

PBDB-T-2F (or PM6) is a wide bandgap polymer donor (n-type semiconductor) containing fluorinated thienyl benzodithiophene (BDT-2F) used in high performance polymer solar cells (PSCs). PBDB-T-2F possesses high crystallinity and strong π-π stacking alignment, which are favourable to charge carrier transport and hence suppress recombination in devices. PBDBT-2F based PSCs were reported to have thickness and area insensitive performance and is a promising candidate for large-scale roll-to-roll manufacturing of high-efficiency polymer solar cells.
For example, recently, new study have shown PBDB-T-2F:IT-4F(Sigma Aldrich Cat. No. 901423) based PSCs yielded an impressive PCE of 13.5% due to the synergistic effect of fluorination on both donor and acceptor, which is among the highest values recorded in the literatures for PSCs to date [1]. The PBDB-T-2F:IT-4F baed PSCs also showed good storage, thermal and illumination stabilities with respect to the efficiency. High efficiency of >11% was maintained for a wide range of film area and thickness. When paired with selenopheno[3,2-b]thiophene-based narrow-bandgap non-fullerene acceptor, an impressive efficiency of 13.3 % was obtained with thickness-insensitive feature.

It has also been previously reported, PBDB-T-2F when paired with narrow band-gap small molecule acceptor 2,2′-((2Z,2′Z)-((4,4,9,9-tetrahexyl-4,9-dihydro-s-indaceno[1,2-b:5,6-b′]dithiophene-2,7-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile (IDIC), the as-cast film (without extra treatments) showed an outstanding power conversion efficiency (PCE) of 11.9%, a record value for as-cast polymer solar cells. Moreover, the performance of PBDB-T-2F:IDIC based devices is insensitive to the active layer thickness (≅95-255 nm) and device area (0.20-0.81 cm2), and thus is a promising candidate for future roll-to-roll mass manufacturing and practical application of highly efficient PSCs.
It has also been reported, PBDB-T-2F possesses a strong absorption in the short wavelength region of 300-685 nm with a large bandgap of 1.80 eV, which is complementary to that of ITIC (1.55 eV) and facilitates achieving high short-circuit current (Jsc) in PSCs. Moreover, PBDB-T-2F shows a deep HOMO level of −5.50 eV, a strong crystallinity and a dominant face on packing, which helps to achieve a high open-circuit voltage (Voc) and fill factor (FF) in PSCs.
PBDB-T-2F can be used as the active semiconductor layer in OFET devices. PBDB-T-2F can serve as the donor material in the photoactive layer of OPV devices. It exhibits abroad absorption spectrum, allowing it to absorb light across a wide range of wavelengths, including visible and near-infrared regions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance nonfullerene polymer solar cells based on a fluorinated wide bandgap copolymer with a high open-circuit voltage of 1.04 V
Wang Y, et al.
Journal of Material Chemistry A, 5(42), 22180-22185 (2017)
Selenopheno[3,2-b]thiophene-Based Narrow-Bandgap Nonfullerene Acceptor Enabling 13.3 % Efficiency for Organic Solar Cells with Thickness-Insensitive Feature
Wang J L, et al.
ACS Energy Letters, 3(12), 2967-2976 (2018)
Qunping Fan et al.
Advanced materials (Deerfield Beach, Fla.), 30(6) (2017-12-14)
In this work, a nonfullerene polymer solar cell (PSC) based on a wide bandgap polymer donor PM6 containing fluorinated thienyl benzodithiophene (BDT-2F) unit and a narrow bandgap small molecule acceptor 2,2'-((2Z,2'Z)-((4,4,9,9-tetrahexyl-4,9-dihydro-s-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile (IDIC) is developed. In addition to matched energy levels
Maojie Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 27(31), 4655-4660 (2015-07-15)
A new copolymer PM6 based on fluorothienyl-substituted benzodithiophene is synthesized and characterized. The inverted polymer solar cells based on PM6 exhibit excellent performance with Voc of 0.98 V and power conversion efficiency (PCE) of 9.2% for a thin-film thickness of
Synergistic effect of fluorination on both donor and acceptor materials for high performance non-fullerene polymer solar cells with 13.5 % efficiency
Qunping F, et al.
Science China: Chemistry, 61(5), 531-537 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service