Skip to Content
Merck
All Photos(1)

Key Documents

900799

Sigma-Aldrich

ITIC

Synonym(s):

2,2′-[[6,6,12,12-Tetrakis(4-hexylphenyl)-6,12-dihydrodithieno[2,3-d:2′,3′-d′]-s-indaceno[1,2-b:5,6-b′]dithiophene-2,8-diyl]bis[methylidyne(3-oxo-1H-indene-2,1(3H)-diylidene)]]bis[propanedinitrile]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C94H82N4O2S4
CAS Number:
Molecular Weight:
1427.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

description

Band gap:1.61eV

Quality Level

Assay

99%

form

solid

Orbital energy

HOMO -5.5 eV 
LUMO -3.89 eV 

SMILES string

O=C1C2=C(C=CC=C2)C(/C1=C/C3=CC(S4)=C(S3)C5=C4C(C=C(C(C6=CC=C(CCCCCC)C=C6)(C7=CC=C(CCCCCC)C=C7)C8=C9SC%10=C8SC(/C=C%11C(C(C=CC=C%12)=C%12C\%11=O)=C(C#N)C#N)=C%10)C9=C%13)=C%13C5(C%14=CC=C(CCCCCC)C=C%14)C%15=CC=C(CCCCCC)C=C%15)=C(C#N)C#N

InChI

1S/C94H82N4O2S4/c1-5-9-13-17-25-59-33-41-65(42-34-59)93(66-43-35-60(36-44-66)26-18-14-10-6-2)79-53-76-80(54-75(79)89-85(93)91-81(103-89)51-69(101-91)49-77-83(63(55-95)56-96)71-29-21-23-31-73(71)87(77)99)94(67-45-37-61(38-46-67)27-19-15-11-7-3,68-47-39-62(

InChI key

HQOWCDPFDSRYRO-VFZXRLAXSA-N

Application

ITIC is a specific non-fullerene acceptor (NFA) molecule used in Organic Photovoltaics(OPVs). ITIC offers potential advantages over traditional fullerene-based acceptors such as include higher Voc (open-circuit voltage), improved stability and broader absorption properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuze Lin et al.
Advanced materials (Deerfield Beach, Fla.), 27(7), 1170-1174 (2015-01-13)
A novel non-fullerene electron acceptor (ITIC) that overcomes some of the shortcomings of fullerene acceptors, for example, weak absorption in the visible spectral region and limited energy-level variability, is designed and synthesized. Fullerene-free polymer solar cells (PSCs) based on the
Wenchao Zhao et al.
Advanced materials (Deerfield Beach, Fla.), 28(23), 4734-4739 (2016-04-12)
A nonfullerene-based polymer solar cell (PSC) that significantly outperforms fullerene-based PSCs with respect to the power-conversion efficiency is demonstrated for the first time. An efficiency of >11%, which is among the top values in the PSC field, and excellent thermal
Yuze Lin et al.
Journal of the American Chemical Society, 138(14), 4955-4961 (2016-03-26)
We develop an efficient fused-ring electron acceptor (ITIC-Th) based on indacenodithieno[3,2-b]thiophene core and thienyl side-chains for organic solar cells (OSCs). Relative to its counterpart with phenyl side-chains (ITIC), ITIC-Th shows lower energy levels (ITIC-Th: HOMO = -5.66 eV, LUMO =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service