901220
EPhos Pd G4
Synonym(s):
[Dicyclohexyl[3-(1-methylethoxy)-2′,4′,6′-tris(1-methylethyl)[1,1′-biphenyl]-2-yl]phosphine-κP](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium
About This Item
Recommended Products
form
powder or crystals
Quality Level
feature
generation 4
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings
functional group
phosphine
SMILES string
[Pd+]c5c(cccc5)c6c(cccc6)NC.P(C4CCCCC4)(C3CCCCC3)c1c(cccc1c2c(cc(cc2C(C)C)C(C)C)C(C)C)OC(C)C.[S](=O)(=O)([O-])C
InChI
1S/C36H55OP.C13H12N.CH4O3S.Pd/c1-24(2)28-22-32(25(3)4)35(33(23-28)26(5)6)31-20-15-21-34(37-27(7)8)36(31)38(29-16-11-9-12-17-29)30-18-13-10-14-19-30;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h15,20-27,29-30H,9-14,16-19H2,1-8H3;2-7,9-10,14H,1H3;1H3,
InChI key
HFYQPKNKOSULDB-UHFFFAOYSA-M
Application
It is a fourth generation (G4) Buchwald precatalyst that is similar to the third generation (G3) precatalysts except that the amino group on the biphenyl backbone is methylated. This modification helps to prevent the limitations of using the third generation precatalysts. It is air, moisture and thermally-stable and shows good solubility in common organic solvents. BrettPhos Pd G4 is highly useful in cross-coupling reactions. Some of its excellent features include lower catalyst loadings, shorter reaction time, and efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service