Skip to Content
Merck
All Photos(1)

Key Documents

799718

Sigma-Aldrich

AlPhos

Synonym(s):

Di-1-adamantyl(4″-butyl-2″,3″,5″,6″-tetrafluoro-2′,4′,6′-triisopropyl-2-methoxy-meta-terphenyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H67F4OP
CAS Number:
Molecular Weight:
815.06
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

Quality Level

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

218-223 °C

functional group

phosphine

storage temp.

−20°C

InChI

1S/C52H67F4OP/c1-9-10-12-38-46(53)48(55)45(49(56)47(38)54)44-40(29(4)5)21-39(28(2)3)43(42(44)30(6)7)37-13-11-14-41(57-8)50(37)58(51-22-31-15-32(23-51)17-33(16-31)24-51)52-25-34-18-35(26-52)20-36(19-34)27-52/h11,13-14,21,28-36H,9-10,12,15-20,22-27H2,1-8H3/t31-,32+,33?,34-,35+,36?,51+,52?,58?

InChI key

ALWIRDZSIXWCBO-VABCSHEKSA-N

Application

AlPhos is a biaryl monophosphine ligand that can be used:
  • In the Pd-catalyzed Buchwald-Hartwig cross-coupling reactions.
  • To synthesize highly regioselective aryl fluorides by Pd-catalyzed fluorination of a variety of activated aryl and heteroaryl triflates and bromides.
  • To prepare aryl thioethers by C–S cross-coupling of thiols with aromatic electrophile in the presence of palladium catalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of Palladium Precatalysts that Efficiently Generate LPd (0) Active Species
Shaughnessy KH
Israel Journal of Chemistry, 60(3-4), 180-194 (2020)
A fluorinated ligand enables room-temperature and regioselective Pd-catalyzed fluorination of aryl triflates and bromides
Sather AC, et al.
Journal of the American Chemical Society, 137(41), 13433-13438 (2015)
In-depth assessment of the palladium-catalyzed fluorination of five-membered heteroaryl bromides
Milner PJ, et al.
Organometallics, 34(19), 4775-4780 (2015)
Monophosphine Ligands Promote Pd-Catalyzed C-S Cross-Coupling Reactions at Room Temperature with Soluble Bases
Xu Jessica, et al.
ACS Catalysis, 9(7), 6461-6466 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service