Skip to Content
Merck
All Photos(1)

Key Documents

900552

Sigma-Aldrich

Graphene

silane modified, avg. no. of layers, 1 ‑ 5

Synonym(s):

Si-G

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiCxHyOz
UNSPSC Code:
12141908
NACRES:
NA.23

Quality Level

form

powder

feature

avg. no. of layers 1 ‑ 5

cation traces

C: >75%
Si: >2.5%

Looking for similar products? Visit Product Comparison Guide

General description

  • Functionalization agent: KH-550.
  • Solubility: Organic solvents.
  • Thickness: 1-5 layers.
  • Lateral size: 0.5-5 μm.

Application

  • Polymer filler to improve thermal, electrical and mechanical properties.
  • Rubber filler to improve mechanical and gas barrier properties.
  • Super hydrophobic surface.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of functionalization of graphene nanoplatelets on the mechanical response of graphene/epoxy composites.
Moghadam A, et al.
Materials & Design, 66, 142-149 (2015)
Oil-absorbent polyurethane sponge coated with KH-570-modified graphene.
Li B, et al.
Journal of Applied Polymer Science, 132(16), 41821/1-41821/7 (2015)
Superhydrophobic Functionalized Graphene Aerogels.
Lin Y, et al.
ACS Applied Materials & Interfaces, 3(7), 2200-2203 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service