Skip to Content
Merck
All Photos(1)

Documents

900551

Sigma-Aldrich

Graphene

amine functionalized, avg. no. of layers, 1 ‑ 5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiCxHyOz
UNSPSC Code:
12141908
NACRES:
NA.23

form

powder

Quality Level

feature

avg. no. of layers 1 ‑ 5

composition

Carbon, >75%
Nitrogen, >2.5%

functional group

amine

Looking for similar products? Visit Product Comparison Guide

General description

  • Functionalization agent: Octadecylamine.
  • Solubility: Organic solvent.
  • Thickness: 1-5 layers.
  • Lateral size : 0.5-5 μm.

Application

  • Polymer filler to improve thermal, electrical and mechanical properties.
  • Rubber filler to improve mechanical and gas barrier properties.
  • Hydrophobic surface.
  • Lubrication.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Rui Wu et al.
Nano letters, 11(12), 5159-5164 (2011-11-19)
We have developed an efficient chemical exfoliation approach for the high-throughput synthesis of solution-processable, high-quality graphene sheets that are noncovalently functionalized by alkylamine. Purely coherent nonlinear optical response of these graphene sheets has been investigated, using near-infrared, visible, and ultraviolet
One-Step Preparation of Form-Stable Phase Change Material through Self-Assembly of Fatty Acid and Graphene.
Akhiani AR, et al.
The Journal of Physical Chemistry C, 119(40), 22787-22796 (2015)
Synthesis and characterization of alkylamine-functionalized graphene for polyolefin-based nanocomposites.
Yang X, et al.
Applied Surface Science, 305, 725-731 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service