Skip to Content
Merck
All Photos(1)

Key Documents

87189

Sigma-Aldrich

1-Tetradecene

≥97.0% (GC)

Synonym(s):

1-Butadecene, n-Tetradec-1-ene, alpha-Tetradecene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CH=CH2
CAS Number:
Molecular Weight:
196.37
Beilstein:
1701156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

6.8 (vs air)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

455 °F

refractive index

n20/D 1.436 (lit.)
n20/D 1.436

bp

251 °C (lit.)

mp

−13-−11 °C (lit.)

density

0.775 g/mL at 25 °C (lit.)

functional group

allyl

SMILES string

CCCCCCCCCCCCC=C

InChI

1S/C14H28/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3H,1,4-14H2,2H3

InChI key

HFDVRLIODXPAHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Tetradecene can be used as a reactant to synthesize:
  • 2-Tetradecanone via PdCl2/CrO3 catalyzed oxidation reaction.[1]
  • (E)-ethyl pentadec-2-enoate, which is used as a key intermediate in the diastereoselective synthesis of (+)-nephrosterinic acid and (+)-protolichesterinic acid via Johnson-Claisen rearrangement.[2]

It can also be used as a monomer unit to synthesize high molecular weight polymer [poly(1- tetradecene)] by titanium complex catalyzed polymerization reaction.[1]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of methyl ketones from terminal olefins using PdCl2/CrO3 system mimicking the Wacker process
Fernandes RA and Bethi V
Tetrahedron, 70(32), 4760-4767 (2014)
Synthesis of ultrahigh molecular weight polymers with low PDIs by polymerizations of 1-decene, 1-dodecene, and 1-tetradecene by Cp* TiMe2 (O-2, 6-iPr2C6H3)-borate catalyst
Nomura K, et al.
Molecules (Basel), 24(8), 1634-1634 (2019)
Diastereoselective synthesis of (+)-nephrosterinic acid and (+)-protolichesterinic acid
Fernandes RA, et al.
Tetrahedron Asymmetry, 23(1), 60-66 (2012)
Marta Krasny et al.
Cell and tissue banking, 16(4), 631-638 (2015-07-15)
Increasingly dental surgeons face the challenge of reconstruction of the height and/or thickness of the alveolar ridge as more and more patients wish to have permanent restoration of their dental defects based on intraosseous implants. Evaluation of human allogeneic bone
Lucas A Lane et al.
The journal of physical chemistry. B, 118(49), 14140-14147 (2014-08-27)
Quantum dots (QDs) offer distinct advantages over organic dyes and fluorescent proteins for biological imaging applications because of their brightness, photostability, and tunability. However, a major limitation is that single QDs emit fluorescent light in an intermittent on-and-off fashion called

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service