Skip to Content
Merck
All Photos(1)

Key Documents

74738

Supelco

1-Octadecene

analytical standard, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15CH=CH2
CAS Number:
Molecular Weight:
252.48
Beilstein:
1761067
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

0.71 (vs air)

Assay

≥99.0% (GC)

autoignition temp.

482 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.444 (lit.)
n20/D 1.445

bp

179 °C/15 mmHg (lit.)
314.4 °C±5.0 °C/760 mmHg (lit.)

mp

14-16 °C (lit.)

transition temp

solidification point 17-19 °C

density

0.789 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCC=C

InChI

1S/C18H36/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2/h3H,1,4-18H2,2H3

InChI key

CCCMONHAUSKTEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Octadecene may be used as a high boiling solvent in the preparation of highly monodispersed silver nanoparticles using silver dodecanoate and dodecanoic acid.
1-Octadecene may be used as a solvent for the synthesis of Co-ferrite nanoparticles (NPs), which involves the decomposition of a reaction mixture containing metal acetylacetonates, hexadecanediol as reducing agent and oleic acid as a surfactant to yield mixed-metal oleate complex, confirmed by fourier transform infrared (FTIR) spectroscopy.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuning the magnetic properties of Co-ferrite nanoparticles through the 1, 2-hexadecanediol concentration in the reaction mixture
Moya C, et al.
Physical Chemistry Chemical Physics, 17(19), 13143-13149 (2015)
An organometallic route to highly monodispersed silver nanoparticles and their application to ink-jet printing
Shim K-I, et al.
Materials Chemistry and Physics, 110(2), 316-321 (2008)
Yaping Du et al.
Journal of the American Chemical Society, 132(5), 1470-1471 (2010-01-19)
Monodisperse Ag(2)S quantum dots (QDs) were synthesized via pyrolysis of Ag(DDTC) in oleic acid, octadecylamine, and 1-octadecene. The uniform alkyl-capped Ag(2)S QDs with a size of 10.2 nm emit near-IR emission at 1058 nm under 785 nm excitation.
Synthesis, shape control, and optical properties of hybrid Au/Fe3O(4) "nanoflowers".
Yanhu Wei et al.
Small (Weinheim an der Bergstrasse, Germany), 4(10), 1635-1639 (2008-07-19)
Zheng Li et al.
Journal of the American Chemical Society, 133(43), 17248-17256 (2011-09-24)
Formation of CdS nanocrystals in the classic approach (with octadecene (ODE) as the solvent and elemental sulfur and cadmium carboxylate as the precursors) was found to be kinetically dependent on reduction of elemental sulfur by ODE, which possessed a critical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service