Skip to Content
Merck
All Photos(2)

Key Documents

806307

Sigma-Aldrich

Hyperbranched G3-PEG20k-OH

Synonym(s):

bis-MPA hyperbranched PEG20k-OH dendrimer, pseudo generation 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C980H1934O498
Molecular Weight:
21687.54
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

Quality Level

mp

56-61 °C

storage temp.

−20°C

General description

  • End Group Functionality: Hydroxyl surface group
  • Generation: Pseudo generation 3
  • No. Surface Groups: 16
  • Calculated Mol. Wt.: 21,688 g/mol
  • Core: Polyethylene Glycol 20,000
  • Branching units: 2,2-bis(methylol)propionic acid (bis-MPA)

Application

Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multifunctional Poly(ethylene glycol): synthesis, characterization, and potential applications of dendritic-linear-dendritic block copolymer hybrids.
Andren O, et al.
Macromolecules, 46(10), 3726-3736 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service