Skip to Content
Merck
All Photos(1)

Key Documents

686573

Sigma-Aldrich

Hyperbranched bis-MPA polyester-64-hydroxyl, generation 4

≥97%

Synonym(s):

ALH-64-OH, Hyperbranched Polymer, Hyperpolymer, PFH-64-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C315H512O189
CAS Number:
Molecular Weight:
7323.32
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

hydroxyl surface groups

Quality Level

Assay

≥97%

form

powder (lyophilized)

mol wt

generation 4

no. Surface Groups

64 (avg.)

storage temp.

2-8°C

SMILES string

CC(CO)(CO)C(=O)OCC(C)(COC(=O)C(C)(CO)CO)C(=O)OCC(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)C(=O)OCC(C)(COC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)C(=O)OCCOCCOCC(COCCOC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)(COCCOC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COCCOC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO

InChI

1S/C315H512O189/c1-255(71-316,72-317)195(380)449-143-291(37,144-450-196(381)256(2,73-318)74-319)231(416)489-175-307(53,176-490-232(417)292(38,145-451-197(382)257(3,75-320)76-321)146-452-198(383)258(4,77-322)78-323)247(432)481-135-287(33,136-482-248(433)308(54,177-491-233(418)293(39,147-453-199(384)259(5,79-324)80-325)148-454-200(385)260(6,81-326)82-327)178-492-234(419)294(40,149-455-201(386)261(7,83-328)84-329)150-456-202(387)262(8,85-330)86-331)227(412)445-67-63-440-61-62-441-191-315(192-442-64-68-446-228(413)288(34,137-483-249(434)309(55,179-493-235(420)295(41,151-457-203(388)263(9,87-332)88-333)152-458-204(389)264(10,89-334)90-335)180-494-236(421)296(42,153-459-205(390)265(11,91-336)92-337)154-460-206(391)266(12,93-338)94-339)138-484-250(435)310(56,181-495-237(422)297(43,155-461-207(392)267(13,95-340)96-341)156-462-208(393)268(14,97-342)98-343)182-496-238(423)298(44,157-463-209(394)269(15,99-344)100-345)158-464-210(395)270(16,101-346)102-347,193-443-65-69-447-229(414)289(35,139-485-251(436)311(57,183-497-239(424)299(45,159-465-211(396)271(17,103-348)104-349)160-466-212(397)272(18,105-350)106-351)184-498-240(425)300(46,161-467-213(398)273(19,107-352)108-353)162-468-214(399)274(20,109-354)110-355)140-486-252(437)312(58,185-499-241(426)301(47,163-469-215(400)275(21,111-356)112-357)164-470-216(401)276(22,113-358)114-359)186-500-242(427)302(48,165-471-217(402)277(23,115-360)116-361)166-472-218(403)278(24,117-362)118-363)194-444-66-70-448-230(415)290(36,141-487-253(438)313(59,187-501-243(428)303(49,167-473-219(404)279(25,119-364)120-365)168-474-220(405)280(26,121-366)122-367)188-502-244(429)304(50,169-475-221(406)281(27,123-368)124-369)170-476-222(407)282(28,125-370)126-371)142-488-254(439)314(60,189-503-245(430)305(51,171-477-223(408)283(29,127-372)128-373)172-478-224(409)284(30,129-374)130-375)190-504-246(431)306(52,173-479-225(410)285(31,131-376)132-377)174-480-226(411)286(32,133-378)134-379/h316-379H,61-194H2,1-60H3

InChI key

GQHXGYXVJHXWBS-UHFFFAOYSA-N

General description

Soluble in polar solvents such as DMSO, and methanol. Emulsifies in water (colloidal system).
calculated mol. wt. 7,323

Other Notes

Bis-MPA =2,2-Bis(hydroxyl-methyl)propionic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Idoia Martín-de-Lucía et al.
Environmental pollution (Barking, Essex : 1987), 249, 581-588 (2019-04-02)
Hyperbranched polymers are nanomaterials belonging to the class of dendritic architectures with increasing applications in many diverse fields. We studied the toxicity of two hyperbranched polymers to the freshwater crustacean Daphnia magna. A hyperbranched hydroxyl-terminated polyester and a commercial hyperbranched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service