Skip to Content
Merck
All Photos(2)

Documents

804193

Sigma-Aldrich

Me3(OMe)tBuXPhos-Pd-G3

Synonym(s):

Me3(OMe)tBuXPhos Pd G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H66NO4PPdS
Molecular Weight:
866.48
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

SMILES string

CC1=C([R])C([R])=C(C)C(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)=C1P(C(C)(C)C)C(C)(C)C.NC3=C(C=CC=C3)C4=C(C=CC=C4)[Pd]OS(C)(=O)=O

General description

Me3(OMe)tBuXPhos-Pd-G-3 is a versatile palladium precatalyst developed by Buchwald group. It is widely employed in various cross-coupling reactions involving the formation of C-C, C-N, C-O, C-F, C-CF3, and C-S bonds. Precatalysts effectively generate active catalytic species for use in various reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service