794473
Ritter Trifluoroiodomethane-TMG Reagent
Synonym(s):
TMG-CF3I, Tetramethylguanidine-trifluoromethyl iodide adduct
About This Item
Recommended Products
form
liquid
Quality Level
reaction suitability
reaction type: C-C Bond Formation
refractive index
n/D 1.461
density
1.495 g/mL at 25 °C
functional group
amine
fluoro
iodo
shipped in
dry ice
storage temp.
2-8°C
SMILES string
FC(F)(F)I.N=C(N(C)C)N(C)C
InChI
1S/C5H13N3.CF3I/c1-7(2)5(6)8(3)4;2-1(3,4)5/h6H,1-4H3;
InChI key
KIOFDNCUJQQUCD-UHFFFAOYSA-N
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service