Skip to Content
Merck
All Photos(3)

Key Documents

241768

Sigma-Aldrich

1,1,3,3-Tetramethylguanidine

99%

Synonym(s):

N,N,N′,N′-Tetramethylguanidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC(=NH)N(CH3)2
CAS Number:
Molecular Weight:
115.18
Beilstein:
969608
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.2 mmHg ( 20 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.469 (lit.)

bp

52-54 °C/11 mmHg (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(=N)N(C)C

InChI

1S/C5H13N3/c1-7(2)5(6)8(3)4/h6H,1-4H3

InChI key

KYVBNYUBXIEUFW-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Related Categories

Application

Base employed in the preparation of alkyl nitriles from alkyl halides and 3′-alkylthymidines from 3′-nitrothymidines.
Promotes the pentavalent bismuth oxidation of primary and secondary alchohols to aldehydes and ketones.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthetic Communications, 23, 2323-2323 (1993)
Tetrahedron Letters, 48, 2885-2885 (2007)
Silvia Tampucci et al.
Pharmaceutics, 12(11) (2020-11-15)
For topical treatment of skin cancer, the design of pH-responsive nanocarriers able to selectively release the drug in the tumor acidic microenvironment represents a reliable option for targeted delivery. In this context, a series of newly synthesized surface-active fatty acid-protic
Jovana Vušurović et al.
ChemistryOpen, 6(6), 739-750 (2017-12-12)
Interactions of ribonucleic acid (RNA) with guanidine and guanidine derivatives are important features in RNA-protein and RNA-drug binding. Here we have investigated noncovalently bound complexes of an 8-nucleotide RNA and six different ligands, all of which have a guanidinium moiety
Tetrahedron, 49, 10061-10061 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service