Skip to Content
Merck
All Photos(1)

Documents

791482

Sigma-Aldrich

EL-HPE high performance electrolyte

Synonym(s):

Greatcell Solar®, EL-HPE, EL-HPE electrolyte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

liquid

Quality Level

color

semi-transparent amber

refractive index

n20/D 1.392

conductivity

16.5-18.5 mS/cm at 20 °C

density

0.875 g/mL at 25 °C

Application

EL-HPE High Performance Electrolyte is a low viscosity, high ionic conductivity liquid electrolyte, and, therefore, provides highest performance.

Due to the low boiling point of the main solvent and the chemistry of the electrolyte mixture; EL-HPE will not provide best long term stability under standard solar conditions and/or under elevated temperature testing

Particularly suitable for glass or metal/glass substrate-based DSC systems.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F

Flash Point(C)

8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service