Skip to Content
Merck
All Photos(1)

Key Documents

78460

Sigma-Aldrich

1,4-Phenylenediamine dihydrochloride

≥99.0%

Synonym(s):

1,4-Benzenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
Beilstein:
3911749
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0%

form

crystals

SMILES string

Cl.Cl.Nc1ccc(N)cc1

InChI

1S/C6H8N2.2ClH/c7-5-1-2-6(8)4-3-5;;/h1-4H,7-8H2;2*1H

InChI key

IYXMNTLBLQNMLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ty J Samo et al.
Environmental microbiology, 20(12), 4385-4400 (2018-07-20)
The surface and surroundings of microalgal cells (phycosphere) are critical interaction zones but have been difficult to functionally interrogate due to methodological limitations. We examined effects of phycosphere-associated bacteria for two biofuel-relevant microalgal species (Phaeodactylum tricornutum and Nannochloropsis salina) using
N Yokdang et al.
British journal of cancer, 104(10), 1628-1640 (2011-04-21)
Human breast carcinoma cells secrete an adenosine 5'-diphosphate transphosphorylase (sNDPK) known to induce endothelial cell tubulogenesis in a P2Y receptor-dependent manner. We examined sNDPK secretion and its effects on human endothelial cells. Nucleoside diphosphate kinase (NDPK) secretion was measured by
Kotaro Morita et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(6), 481-485 (2015-06-13)
A photoluminescent detection method for nitrite with high selectivity and sensitivity using carbon nanodots (CNDs) is demonstrated. The selectivity of nitrite is accomplished by a highly specific diazotization reaction between nitrite and p-phenylenediamine (p-PDA). In the presence of nitrite, p-PDA
Giovanni Rizzo et al.
PloS one, 7(11), e50230-e50230 (2012-12-05)
Leber's hereditary optic neuropathy (LHON) is characterized by retinal ganglion cell (RGC) degeneration with the preferential involvement of those forming the papillomacular bundle. The optic nerve is considered the main pathological target for LHON. Our aim was to investigate the
Laura M Pot et al.
Contact dermatitis, 68(4), 193-207 (2013-03-21)
Although p-phenylenediamine (PPD) has been recognized as an extreme sensitizer for many years, the exact mechanism of sensitization has not been elucidated yet. Penetration and the ability to bind to proteins are the first two hurdles that an allergen has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service