Skip to Content
Merck
All Photos(1)

Key Documents

78450

Sigma-Aldrich

m-Phenylenediamine dihydrochloride

≥99.0% (AT)

Synonym(s):

1,3-Benzenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2 · 2HCl
CAS Number:
Molecular Weight:
181.06
Beilstein:
3911748
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (AT)

form

powder

SMILES string

Cl.Cl.Nc1cccc(N)c1

InChI

1S/C6H8N2.2ClH/c7-5-2-1-3-6(8)4-5;;/h1-4H,7-8H2;2*1H

InChI key

SVTOYMIYCMHPIV-UHFFFAOYSA-N

Application

m-Phenylenediamine dihydrochloride is used in the synthesis of bis(thiazolidone) and bis(thiohydantoin) derivatives. It can be used to increase the pore size in the fabrication of poly(aniline-co-1,3-phenylenediamine) cryogels.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SYNTHESIS OF BIS (THIAZOLIDONE) AND BIS (THIOHYDANTOIN) AND THEIR DERIVATIVES FROM M-PHENYLENEDIAMINE.
ACHARY T E, et al.
Chemischer Informationsdienst, 7(16) (1976)
Effect of 1, 3-phenylenediamine concentration on the properties of poly (aniline-co-1, 3-phenylenediamine) cryogels.
Milakin K A, et al.
Materials Letters (2018)
Ariana Q Farrand et al.
Brain research, 1651, 88-94 (2016-10-30)
Mitochondrial dysfunction has been implicated in the degeneration of dopamine (DA) neurons in Parkinson's disease (PD). In addition, animal models of PD utilizing neurotoxins, such as 6-hydroxydopamine and 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine, have shown that these toxins disrupt mitochondrial respiration by targeting complex
D M Bortz et al.
Neuropharmacology, 121, 69-78 (2017-04-19)
Cognitive deficits represent core symptoms in schizophrenia (SZ) and predict patient outcome; however, they remain poorly treated by current antipsychotic drugs. Elevated levels of the endogenous alpha7 nicotinic receptor negative allosteric modulator and NMDA receptor antagonist, kynurenic acid (KYNA), are
N Yokdang et al.
British journal of cancer, 104(10), 1628-1640 (2011-04-21)
Human breast carcinoma cells secrete an adenosine 5'-diphosphate transphosphorylase (sNDPK) known to induce endothelial cell tubulogenesis in a P2Y receptor-dependent manner. We examined sNDPK secretion and its effects on human endothelial cells. Nucleoside diphosphate kinase (NDPK) secretion was measured by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service