Skip to Content
Merck
All Photos(1)

Key Documents

777293

Sigma-Aldrich

2-{[7-(5-N,N-Ditolylaminothiophen-2-yl)-2,1,3-benzothiadiazol-4-yl]methylene}malononitrile

99% (HPLC)

Synonym(s):

2-[[7-[5-[Bis(4-methylphenyl)amino]-2-thienyl]-2,1,3-benzothiadiazol-4-yl]methylene]propanedinitrile, DTDCTB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H19N5S2
CAS Number:
Molecular Weight:
489.61
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% (HPLC)

form

powder

mp

230-235 °C

transition temp

Tm 233 °C

λmax

662-664 nm in dichloromethane

SMILES string

CC(C=C1)=CC=C1N(C2=CC=C(C)C=C2)C3=CC=C(C4=CC=C(C=C(C#N)C#N)C5=NSN=C54)S3

InChI

1S/C28H19N5S2/c1-18-3-8-22(9-4-18)33(23-10-5-19(2)6-11-23)26-14-13-25(34-26)24-12-7-21(15-20(16-29)17-30)27-28(24)32-35-31-27/h3-15H,1-2H3

InChI key

BCJCBXQJAANTJL-UHFFFAOYSA-N

General description

2-{[7-(5-N,N-Ditolylaminothiophen-2-yl)-2,1,3-benzothiadiazol-4-yl]methylene}malononitrile (DTDCTB) is a conducting polymer that can be used as a donor molecule. It is majorly used in the development of organic electronics.
Device structure:
MoO3 (30nm) / DTDCTB (7nm) / DTDCTB:C60/C70 (40nm) / C60/C70 (7nm) / BCP (10nm) / Ag

Device performance:
  • JSC = 14.68 mA/cm2
  • VOC = 0.8 V
  • FF = 0.5
  • PCE = 5.81%

Application

A vacuum-deposited organic solar cell employing this novel donor-acceptor-acceptor (D-A-A) donor molecule; DTDCTB; combined with the electron acceptor C60/ C70 achieved a record-high power conversion efficiency (PCE) of 5.81%.
DTDCTB is a π-conjugating polymer that can be used in the fabrication of organic solar cells (OSCs) and organic photovoltaic cells (OPVs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of the charge-transfer states in the ultrafast excitonic dynamics of the DTDCTB dimers embedded in a crystal environment.
Jiang S, et al.
Chemical Physics, 515, 603-613 (2018)
Highly efficient bifacial transparent organic solar cells with power conversion efficiency greater than 3% and transparency of 50%.
Lin H, et al.
Organic Electronics, 13(9), 1722-1728 (2012)
Deposition growth and morphologies of C60 on DTDCTB surfaces: An atomistic insight into the integrated impact of surface stability, landscape, and molecular orientation.
Han G, et al.
Advanced Materials Interfaces, 2(17), 1500329-1500329 (2015)
An effective bilayer cathode buffer for highly efficient small molecule organic solar cells.
Lin H, et al.
Organic Electronics, 13(10), 1925-1929 (2012)
Li-Yen Lin et al.
Journal of the American Chemical Society, 133(40), 15822-15825 (2011-09-13)
A novel donor-acceptor-acceptor (D-A-A) donor molecule, DTDCTB, in which an electron-donating ditolylaminothienyl moiety and an electron-withdrawing dicyanovinylene moiety are bridged by another electron-accepting 2,1,3-benzothiadiazole block, has been synthesized and characterized. A vacuum-deposited organic solar cell employing DTDCTB combined with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service