Skip to Content
Merck
All Photos(2)

Documents

777242

Sigma-Aldrich

Silica, mesostructured

SBA-15, 99% trace metals basis

Synonym(s):

Silica, Porous Silica, nanostructured silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% trace metals basis

form

solid

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

SBA-15 is a well studied and highly stable mesoporous silica with applications in drug delivery; enzyme immobilization; catalysis; and as templating agents.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

María Vallet-Regí et al.
Angewandte Chemie (International ed. in English), 46(40), 7548-7558 (2007-09-15)
Research on mesoporous materials for biomedical purposes has experienced an outstanding increase during recent years. Since 2001, when MCM-41 was first proposed as drug-delivery system, silica-based materials, such as SBA-15 or MCM-48, and some metal-organic frameworks have been discussed as
S H Joo et al.
Nature, 412(6843), 169-172 (2001-07-13)
Nanostructured carbon materials are potentially of great technological interest for the development of electronic, catalytic and hydrogen-storage systems. Here we describe a general strategy for the synthesis of highly ordered, rigid arrays of nanoporous carbon having uniform but tunable diameters
Cathleen M Crudden et al.
Journal of the American Chemical Society, 127(28), 10045-10050 (2005-07-14)
The functionalization of SBA-15 with mercaptopropyl trimethoxysilane leads to a material capable of absorbing Pd from organic and aqueous solutions. The resulting Pd-loaded material acts as a catalyst for the Suzuki-Miyaura and Mizoroki-Heck coupling reactions. Leaching studies show that the
Catalytic Activity in Organic Solvents and Stability of Immobilized Enzymes Depend on the Pore Size and Surface Characteristics of Mesoporous Silica
Takahashi; H.;
Chemistry of Materials, 12(11), 3301-3305 (2000)
Keren B Turton et al.
PloS one, 10(5), e0127243-e0127243 (2015-05-20)
Signal transducer and activator of transcription 3 (STAT3) is a key mediator of leukocyte differentiation and proliferation. The 3' end of STAT3 transcripts is subject to two alternative splicing events. One results in either full-length STAT3α or in STAT3β, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service