Skip to Content
Merck
All Photos(3)

Documents

643645

Sigma-Aldrich

Silica, mesostructured

MCM-41 type (hexagonal)

Synonym(s):

Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

surface area

~1000 m2/g , BET

unit cell size

4.5-4.8 nm

pore size

0.98 cm3/g pore volume
2.1-2.7 nm pore size

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

bulk density

0.34 g/mL

application(s)

battery manufacturing

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica, mesostructured (MCM-41) is a part of low-density organized mesoporous silicate system which has a 2D hexagonal phase, prepared in the presence of quaternary ammonium halide surfactant. It has ordered and large pores with diameter in the range 1.5-10 nm and specific surface area of 1500 m2/g.

Application

MCM-41 may be used as a base material that can be assembled with platinum complexes to develop luminescent oxygen sensing materials. It may also be used as an adsorbent in CO2 capture application. Surface functionalized MCM-41 may be used as a catalyst in the synthesis of bisphenol A.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chiral mesostructured silica nanofibers of MCM-41
Wang B, et al.
Angewandte Chemie (International ed. in English), 45(13), 2088-2090 (2006)
Adsorption of CO2-containing gas mixtures over amine-bearing pore-expanded MCM-41 silica: application for gas purification
Belmabkhout Y, et al.
Industrial & Engineering Chemistry Research, 49(1), 359-365 (2009)
In situ investigations on cetyltrimethylammonium surfactant/silicate systems, precursors of organized mesoporous MCM-41-type siliceous materials
Frasch J, et al.
Langmuir, 16(23), 9049-9057 (2000)
Oxygen sensing materials based on mesoporous silica MCM-41 and Pt (II)-porphyrin complexes
Zhang H, et al.
Journal of Materials Chemistry, 15(31), 3181-3186 (2005)
Effect of surface functionalization of MCM-41-type mesoporous silica nanoparticles on the endocytosis by human cancer cells
Slowing I, et al.
Journal of the American Chemical Society, 128(46), 14792-14793 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service