Skip to Content
Merck
All Photos(1)

Documents

773972

Sigma-Aldrich

Yttrium sputtering target

diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis

Synonym(s):

Yttrium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

solid

reaction suitability

core: yttrium

resistivity

57 μΩ-cm, 20°C

diam. × thickness

2.00 in. × 0.25 in.

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solid Oxide Fuel cells operating at temperatures below 800 C (also known as intermediate temperature solid oxide fuel cell, IT-SOFC) are currently the topic of much research and development owing to the high degradation rates and materials costs incurred for SOFC operating at temperatures above 900 C. Thin films of electrode and electrolyte layers is one of the ways to achieve high performances in IT-SOFC.
Yttrium sputtering target can be used for physical vapor deposition of thin films of yttria stabilized zirconia layers for IT-SOFC. Yttrium containing thin films also find applications as thermal barrier and protective coatings, example in thermoelectric devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Blanco-Canosa et al.
Physical review letters, 110(18), 187001-187001 (2013-05-21)
We use resonant x-ray scattering to determine the momentum-dependent charge correlations in YBa(2)Cu(3) O(6.55) samples with highly ordered chain arrays of oxygen acceptors (ortho-II structure). The results reveal nearly critical, biaxial charge density wave (CDW) correlations at in-plane wave vectors
Yttrium-catalyzed addition of benzylic C-H bonds of alkyl pyridines to olefins.
Bing-Tao Guan et al.
Angewandte Chemie (International ed. in English), 52(16), 4418-4421 (2013-03-21)
José Renato Queiroz et al.
The journal of adhesive dentistry, 15(2), 151-159 (2013-03-28)
To compare the effect of silica (Si)-based nano-coating deposited by reactive magnetron sputtering (RMP) with that of conventional surface conditioning using metal/zirconia primer alone or after air-particle abrasion on the adhesion of resin cements to zirconia ceramic. Two hundred forty
Masahiko Ooe et al.
Aesthetic plastic surgery, 37(2), 424-433 (2013-02-12)
Skin wrinkles are one of the most cosmetically concerning signs of aging for women, and improvements in the visual effect of wrinkles become a matter of concern that has an impact on the quality of life. Although various wrinkle treatments
Gitanjal Deka et al.
Journal of biomedical optics, 19(1), 011012-011012 (2013-08-21)
Cellular micropattering has been increasingly adopted in quantitative biological experiments. A Q-switched pulsed neodymium-doped yttrium ortho-vanadate (Nd∶YVO4) laser directed in-situ microfabrication technique for cell patterning is presented. A platform is designed uniquely to achieve laser ablation. The platform is comprised

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service