Skip to Content
Merck
All Photos(1)

Documents

746959

Sigma-Aldrich

Ethenesulfonyl fluoride

95%

Synonym(s):

ESF, Vinyl sulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3FO2S
CAS Number:
Molecular Weight:
110.11
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.385

density

1.328 g/mL at 25 °C

SMILES string

O=S(F)(C=C)=O

InChI

1S/C2H3FO2S/c1-2-6(3,4)5/h2H,1H2

InChI key

BYPHZHGVWNKAFC-UHFFFAOYSA-N

General description

Ethenesulfonylfluoride is a Michael acceptor used in dyestuffs, functional materials,photoresist materials, lubricating oil additives, and medicinal chemistry. Itexhibits extraordinary reactivity in SuFEx click chemistry and organicsynthesis.

Application

Ethenesulfonyl fluoride can be used as a reactant to synthesize:
  • β-Arylethenesulfonyl fluorides via palladium(II) acetate-catalyzed Heck-Matsuda reaction with arenediazonium tetrafluoroborates.
  • Bisalkylsulfonyl fluoride(BSF) monomers via Michael addition reaction with amines/anilines. BSF monomers are applicable in the synthesis of polysulfonates.
  • Cyclobutane-fused pyridinyl sulfonyl fluorides by photocatalytic [2 + 2] cycloaddition with pyridones or isoquinolones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno A da S Dantas et al.
Revista brasileira de psiquiatria (Sao Paulo, Brazil : 1999), 42(2), 201-208 (2019-12-12)
To evaluate the impact of multidimensional interventions on quality of life (QoL) and depressive symptoms in Brazilian older adults living in the community. Longitudinal, quasi-experimental study of older adults receiving conventional primary health care (PHC). The interventions were designed in
Markus Wolfien et al.
EBioMedicine, 57, 102862-102862 (2020-07-07)
Bone marrow stem cell clonal dysfunction by somatic mutation is suspected to affect post-infarction myocardial regeneration after coronary bypass surgery (CABG). Transcriptome and variant expression analysis was studied in the phase 3 PERFECT trial post myocardial infarction CABG and CD133+
Christin Völkner et al.
Stem cell research, 50, 102127-102127 (2020-12-29)
Niemann-Pick disease Type C (NPC) is a rare progressive neurodegenerative disorder with an incidence of 1:120,000 caused by mutations in the NPC1 or NPC2 gene leading to a massive cholesterol accumulation. Here, we describe the generation of induced pluripotent stem
Jiranuwat Sapudom et al.
Scientific reports, 7(1), 14135-14135 (2017-10-28)
Live cell imaging enables an observation of cell behavior over a period of time and is a growing field in modern cell biology. Quantitative analysis of the spatio-temporal dynamics of heterogeneous cell populations in three-dimensional (3D) microenvironments contributes a better
Monika Zouharova et al.
International journal of molecular sciences, 20(18) (2019-09-12)
Transient receptor potential (TRPs) channels are crucial downstream targets of calcium signalling cascades. They can be modulated either by calcium itself and/or by calcium-binding proteins (CBPs). Intracellular messengers usually interact with binding domains present at the most variable TRP regions-N-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service