Skip to Content
Merck
All Photos(1)

Key Documents

767344

Sigma-Aldrich

Poly(D,L-lactide)

average Mn 20,000 (NMR), PDI ≤1.3

Synonym(s):

PDLA, PLA, polyDL-lactide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

crystals

mol wt

average Mn 20,000 (NMR)

degradation timeframe

<6 months

mp

320-350 °C

PDI

≤1.3

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Used in Drug Delivery including coating nanoparticles. Can be end-group functionalizated or used as a macroinitiator/block precursor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Shape Amphiphiles Based on Functional Polyhedral Oligomeric Silsesquioxane End-Capped Poly(L-Lactide) with Diverse Head Surface Chemistry
Zhang W B, et al.
Macromolecules, 44(8), 2589-2596 (2011)
Poly(lactide)-block-Poly(HEMA) Block Copolymers: An Orthogonal One-Pot Combination of ROP and ATRP, Using a Bifunctional Initiator
Wolf F F, et al.
Macromolecules, 42(15), 5622-5628 (2009)
Preparation of Protein-Loaded Poly(L-Lactide) Microspheres by Solution-Enhanced Dispersion by Supercritical CO2
Chen A Z, et al.
Journal of Biomimetics, Biomaterials, and Tissue Engineering, 11, 93-100 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service