805734
Poly(D,L-lactide)
acid terminated, average Mw 15000
Synonym(s):
PDLLA, Polylactide, lactide polymer
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(C3H4O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
Quality Level
form
solid
mol wt
average Mw 15000
degradation timeframe
<6 months
storage temp.
2-8°C
SMILES string
O1C(C(=O)OC(C1=O)C)C
InChI
1S/C6H8O4/c1-3-5(7)10-4(2)6(8)9-3/h3-4H,1-2H3
InChI key
JJTUDXZGHPGLLC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
May be used to make drug delivery materials for controlled release.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service