Skip to Content
Merck
All Photos(2)

Key Documents

766038

Sigma-Aldrich

N,N′-Bis[2-(2-tert-butyldimethylsilyloxyethoxy)ethyl]-3,4,9,10-perylenetetracarboxylic diimide

97%

Synonym(s):

2,9-Bis[2-[2-[[(1,1-dimethylethyl)dimethylsilyl]oxy]ethoxy]ethyl]anthra[2,1,9-def:6,5,10-d′e′f′] diisoquinoline-1,3,8,10(2H,9H)-tetrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H54N2O8Si2
CAS Number:
Molecular Weight:
795.08
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

357-362 °C

storage temp.

2-8°C

SMILES string

O=C(N(CCOCCO[Si](C)(C)C(C)(C)C)C1=O)C2=CC=C3C4=CC=C5C6=C(C(N(CCOCCO[Si](C)(C(C)(C)C)C)C5=O)=O)C=CC(C7=C3C2=C1C=C7)=C46

InChI

1S/C44H54N2O8Si2/c1-43(2,3)55(7,8)53-25-23-51-21-19-45-39(47)31-15-11-27-29-13-17-33-38-34(18-14-30(36(29)38)28-12-16-32(40(45)48)37(31)35(27)28)42(50)46(41(33)49)20-22-52-24-26-54-56(9,10)44(4,5)6/h11-18H,19-26H2,1-10H3

InChI key

MQIYPIYWJFMSSJ-UHFFFAOYSA-N

Application

Planar perylene- and naphthalene-based diimide linkers can be employed to tether the Watson-
Crick and the Hoogsteen strands of a DNA triplex, thus providing conjugates capable of targeting singlestranded nucleic acids with the formation of hairpin triplexes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan Bevers, et al.
Journal of the American Chemical Society, 122, 5905-5915 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service