Skip to Content
Merck
All Photos(5)

Documents

754242

Sigma-Aldrich

Tetrahydroxydiboron

95%

Synonym(s):

BBA, Bis-Boric acid, Diboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B2H4O4
CAS Number:
Molecular Weight:
89.65
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

>385 °C

SMILES string

OB(O)B(O)O

InChI

1S/B2H4O4/c3-1(4)2(5)6/h3-6H

InChI key

SKOWZLGOFVSKLB-UHFFFAOYSA-N

Application

Tetrahydroxydiboron (BBA) is an efficient borylating agent that can be used to borylate a wide variety of aryl and heteroaryl substrates with low Pd- and Ni-catalyst loads.

Other reactions employing BBA as a reagent:
  • Palladium-catalyzed boronation of vinyl cyclopropane, vinyl aziridine, and allyl acetate substrates.
  • To facilitate catalytic transfer hydrogenations of simple alkenes and alkynes.
  • Selective reducing agent for in situ N-oxide reduction of pyridine-N-oxides.
  • As a substitute to bis(pinacolato) diboron for Miyaura borylation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and Selective in situ Reduction of Pyridine-N-oxides with Tetrahydroxydiboron.
Londregan A T
Synlett, 24(20), 2695-2700 (2013)
Scope of the Palladium-Catalyzed Aryl Borylation Utilizing Bis-Boronic Acid.
Molander GA, et al.
Journal of the American Chemical Society, 134, 11667-11673 (2012)
Nickel-catalyzed borylation of halides and pseudohalides with tetrahydroxydiboron [B2 (OH) 4].
Molander G A, et al.
The Journal of Organic Chemistry, 78(13), 6427-6439 (2013)
Palladium-catalyzed, direct boronic acid synthesis from aryl chlorides: a simplified route to diverse boronate ester derivatives.
Molander G A, et al.
Journal of the American Chemical Society, 132(50), 17701-17703 (2010)
Palladium pincer complex catalyzed substitution of vinyl cyclopropanes, vinyl aziridines, and allyl acetates with tetrahydroxydiboron. An efficient route to functionalized allylboronic acids and potassium trifluoro (allyl) borates.
Sebelius S et al.
Journal of the American Chemical Society, 127(30), 10478-10479 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service