518808
Bis(neopentyl glycolato)diboron
96%
Synonym(s):
5,5,5′,5′-Tetramethyl-2,2′-bi-1,3,2-dioxaborinane
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H20B2O4
CAS Number:
Molecular Weight:
225.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
96%
mp
180.5-184.5 °C (lit.)
SMILES string
CC1(C)COB(OC1)B2OCC(C)(C)CO2
InChI
1S/C10H20B2O4/c1-9(2)5-13-11(14-6-9)12-15-7-10(3,4)8-16-12/h5-8H2,1-4H3
InChI key
MDNDJMCSXOXBFZ-UHFFFAOYSA-N
Application
Reagent to prepare boronic acid esters.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
George W Kabalka et al.
Nuclear medicine and biology, 30(4), 369-372 (2003-05-28)
A direct radioiodination of (Z)-vinylboronic acid esters to the corresponding vinyl iodides using Na(123)I and chloramine-T is described. The boronates were prepared from vinyl iodides via palladium coupling reactions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service