All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H4F2O4S2
CAS Number:
Molecular Weight:
242.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
solid
reaction suitability
reaction type: click chemistry
mp
37-42 °C
SMILES string
FS(C1=CC(S(F)(=O)=O)=CC=C1)(=O)=O
InChI
1S/C6H4F2O4S2/c7-13(9,10)5-2-1-3-6(4-5)14(8,11)12/h1-4H
InChI key
VCINFPPPNNZOAD-UHFFFAOYSA-N
Application
Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service