735892
Poly(styrene)-block-poly(acrylic acid), azide terminated
average Mn 8,700
Synonym(s):
PS-b-PAA, Polystyrene-polyacrylic acid polymersome forming polymer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH(C6H5)[CH2CH(C6H5)]x [CH2CH(CO2H)]y N3
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
solid
mol wt
Mn 1,600-1,950 (poly(acrylic acid))
Mn 6,500-7,000 (polystyrene)
Mn 8,100-9,100
average Mn 8,700
mp
192-197 °C
Mw/Mn
1.2
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service