All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5HCl3N2O
CAS Number:
Molecular Weight:
211.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.586
density
1.645 g/mL at 25 °C
functional group
acyl chloride
chloro
SMILES string
ClC(=O)c1cnc(Cl)nc1Cl
InChI
1S/C5HCl3N2O/c6-3-2(4(7)11)1-9-5(8)10-3/h1H
InChI key
DZTIFMWYYHCREC-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
199.9 °F
Flash Point(C)
93.3 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service