730149
N-(3-Methoxypropyl)acrylamide
contains MEHQ as inhibitor, 95%
Synonym(s):
MPAM, NMPA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
95%
form
liquid
contains
MEHQ as inhibitor
refractive index
n20/D 1.475
density
1.023 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
COCCCNC(=O)C=C
InChI
1S/C7H13NO2/c1-3-7(9)8-5-4-6-10-2/h3H,1,4-6H2,2H3,(H,8,9)
InChI key
RWJGITGQDQSWJG-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chiara Russo et al.
Archives of toxicology, 93(1), 179-188 (2018-10-21)
Cannabidiol (CBD) and cannabidivarin (CBDV) are natural cannabinoids which are consumed in increasing amounts worldwide in cannabis extracts, as they prevent epilepsy, anxiety, and seizures. It was claimed that they may be useful in cancer therapy and have anti-inflammatory properties.
Giampaolo Ricci et al.
PloS one, 6(2), e16912-e16912 (2011-03-02)
Genome-wide association studies of pooled DNA samples were shown to be a valuable tool to identify candidate SNPs associated to a phenotype. No such study was up to now applied to childhood allergic asthma, even if the very high complexity
Ramachandran Samivel et al.
Oxidative medicine and cellular longevity, 2020, 1246510-1246510 (2020-07-04)
Ultraviolet radiation is an environmental carcinogenic agent that enhances inflammation and immunological reactions in the exposed human skin cells leading to oxidative photoaging of the epidermal and dermal segment. In the present study, we investigated the protective role of ursolic
Mirta Milić et al.
Arhiv za higijenu rada i toksikologiju, 69(2), 154-168 (2018-07-11)
In this 28 day-study, we evaluated the effects of herbicide glyphosate administered by gavage to Wistar rats at daily doses equivalent to 0.1 of the acceptable operator exposure level (AOEL), 0.5 of the consumer acceptable daily intake (ADI), 1.75 (corresponding
Jessica Timóteo Jeremias et al.
International braz j urol : official journal of the Brazilian Society of Urology, 47(2), 275-283 (2020-11-05)
Sperm DNA fragmentation is a major cellular mechanism underlying varicocele-related male infertility. However, the type of DNA fragmentation - whether oxidative or of another nature - remains unknown. Thus, the aim of this study was to evaluate single- and double-stranded
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service