Skip to Content
Merck
All Photos(2)

Key Documents

728365

Sigma-Aldrich

1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one

≥98.0% (AT)

Synonym(s):

1-{[Tris-(1-methylethyl)silyl]ethynyl]}-1,2-benziodoxol-3(1H)-one, TIPS-EBX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25IO2Si
CAS Number:
Molecular Weight:
428.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (AT)

form

crystals

reaction suitability

reaction type: C-C Bond Formation

functional group

iodo

SMILES string

CC(C)[Si](C#C[I]1OC(=O)c2ccccc12)(C(C)C)C(C)C

InChI

1S/C18H25IO2Si/c1-13(2)22(14(3)4,15(5)6)12-11-19-17-10-8-7-9-16(17)18(20)21-19/h7-10,13-15H,1-6H3

InChI key

NTHGHMCOPNSZIR-UHFFFAOYSA-N

Application

Introduction of silylacetylenes

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct alkynylation of thiophenes: cooperative activation of TIPS-EBX with gold and Brønsted acids.
Jonathan P Brand et al.
Angewandte Chemie (International ed. in English), 49(40), 7304-7307 (2010-08-24)
Direct alkynylation of indole and pyrrole heterocycles.
Jonathan P Brand et al.
Angewandte Chemie (International ed. in English), 48(49), 9346-9349 (2009-11-07)
Stefano Nicolai et al.
Organic letters, 12(2), 384-387 (2009-12-17)
The first example of intramolecular oxyalkynylation of nonactivated alkenes using oxidative Pd chemistry is reported. Both phenol and aromatic or aliphatic acid derivatives could be used under operator-friendly conditions (room temperature, technical solvents, under air). The discovery of the superiority

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service