Skip to Content
Merck
All Photos(3)

Key Documents

727881

Sigma-Aldrich

trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile

≥98%

Synonym(s):

DCTB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2
CAS Number:
Molecular Weight:
250.34
Beilstein:
8913051
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%
≥98.0% (HPLC)

form

powder

SMILES string

CC(=C/c1ccc(cc1)C(C)(C)C)\C=C(\C#N)C#N

InChI

1S/C17H18N2/c1-13(10-15(11-18)12-19)9-14-5-7-16(8-6-14)17(2,3)4/h5-10H,1-4H3/b13-9+

InChI key

OIASAVWSBWJWBR-UKTHLTGXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Soluble Microporous Ladder Polymers Formed by Stepwise Nucleophilic Substitution of Octafluorocyclopentene: This study utilizes trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile in the context of creating soluble microporous polymers, particularly highlighting its utility in enhancing the properties of these materials. The detailed characterization and application in ionization spectra are discussed (Krishnan and Parthiban).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aisling Byrne et al.
Dalton transactions (Cambridge, England : 2003), 44(32), 14323-14332 (2015-07-23)
A first investigation into the application of a luminescent osmium(ii) bipyridine complex to live cell imaging is presented. Osmium(ii) (bis-2,2-bipyridyl)-2(4-carboxylphenyl) imidazo[4,5f][1,10]phenanthroline was prepared and conjugated to octaarginine, a cell penetrating peptide. The photophysics, cell uptake and cytotoxicity of this osmium
Zhihao Huang et al.
Nature communications, 10(1), 1918-1918 (2019-04-25)
Digital polymers with precisely ordered units acting as the coded 0- or 1-bit, are introduced as a promising option for molecular data storage. However, the pursuit of better performance in terms of high storage capacity and useful functions never stops.
Paul A Scattergood et al.
Dalton transactions (Cambridge, England : 2003), 44(26), 11705-11716 (2015-02-17)
The intense absorption in the red part of the visible range, and the presence of a lowest charge-transfer excited state, render Platinum(II) diimine catecholates potentially promising candidates for light-driven applications. Here, we test their potential as sensitisers in dye-sensitised solar
Baowei Zhang et al.
Nature communications, 9(1), 2499-2499 (2018-06-29)
Structural isomerism of colloidal semiconductor nanocrystals has been largely unexplored. Here, we report one pair of structural isomers identified for colloidal nanocrystals which exhibit thermally-induced reversible transformations behaving like molecular isomerization. The two isomers are CdS magic-size clusters with sharp
Matthew B Baker et al.
Nature communications, 6, 6234-6234 (2015-02-24)
The rational design of supramolecular polymers in water is imperative for their widespread use, but the design principles for these systems are not well understood. Herein, we employ a multi-scale (spatial and temporal) approach to differentiate two analogous water-soluble supramolecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service