Skip to Content
Merck
All Photos(1)

Key Documents

688495

Sigma-Aldrich

(S)-4-tert-Butyl-2-[2-(diphenylphosphino)phenyl]-2-oxazoline

97%

Synonym(s):

(4S)-tert-Butyl-2-[2-(diphenylphosphino)phenyl]-4,5-dihydrooxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H26NOP
CAS Number:
Molecular Weight:
387.45
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]22/D -57.0°, c = 1 in chloroform

mp

111-116 °C

storage temp.

−20°C

SMILES string

CC(C)(C)[C@@H]1N=C(C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4)OC1

InChI

1S/C25H26NOP/c1-25(2,3)23-18-27-24(26-23)21-16-10-11-17-22(21)28(19-12-6-4-7-13-19)20-14-8-5-9-15-20/h4-17,23H,18H2,1-3H3/t23-/m1/s1

InChI key

DMOLTNKQLUAXPI-HSZRJFAPSA-N

General description

The product is a phosphinooxazoline (PHOX) ligand introduced by Pfaltz and coworkers. These non-symmetrical modular P,N-ligands are particularly useful in cases where double bond migration leads to undesired products or mixtures of isomers.

Application

(S)-4-tert-Butyl-2-[2-(diphenylphosphino)phenyl]-2-oxazoline can be used as a ligand for enantioselective Heck reaction without any C=C double bond migration byproduct. It can also be used in a rhodium(I)-catalyzed enantioselective desymmetrization reaction of meso-3,5-dimethyl glutaric anhydride, to form substituted syn-deoxypolypropionate fragments in a single transformation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PHOX
Aldrich Chemfiles, 8(2) null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service