Skip to Content
Merck
All Photos(2)

Key Documents

717223

Sigma-Aldrich

11-(1H-pyrrol-1-yl)undecane-1-thiol

96%

Synonym(s):

PUT, 1H-Pyrrole-1-undecanethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H27NS
CAS Number:
Molecular Weight:
253.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

liquid

refractive index

n20/D 1.506

density

0.928 g/mL at 25 °C

SMILES string

SCCCCCCCCCCCn1cccc1

InChI

1S/C15H27NS/c17-15-11-7-5-3-1-2-4-6-8-12-16-13-9-10-14-16/h9-10,13-14,17H,1-8,11-12,15H2

InChI key

JJDNIFVXOIKYCT-UHFFFAOYSA-N

General description

11-(1H-pyrrol-1-yl)undecane-1-thiol (PUT) is a derivative of pyrrole that forms a self-assembled monolayer (SAM) with terminal pyrrol groups. The pyrrol groups facilitate the formation of a conductive monolayer on a variety of surfaces.

Application

PUT can be used as a SAM on the gold surface that may be utilized in micro-contact printing technique.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristel Panksep et al.
Toxins, 12(4) (2020-04-01)
Global warming, paired with eutrophication processes, is shifting phytoplankton communities towards the dominance of bloom-forming and potentially toxic cyanobacteria. The ecosystems of shallow lakes are especially vulnerable to these changes. Traditional monitoring via microscopy is not able to quantify the
A polypyrrole/polymethylene pattern on gold using a micro-contact printing technique.
Grace AN and Pandian K
Journal of Solid State Electrochemistry, 7(5), 296-300 (2003)
Lisa B Weiss et al.
The journal of physical chemistry. B, 122(22), 5940-5950 (2018-05-16)
We have used nonequilibrium molecular dynamics to simulate the flow of water molecules around a charged nanoparticle described at the atomic scale. These nonequilibrium simulations allowed us to compute the friction coefficient of the nanoparticle and then to deduce its
Ana Weidenauer et al.
Translational psychiatry, 10(1), 2-2 (2020-02-19)
Schizophrenia is characterized by increased behavioral and neurochemical responses to dopamine-releasing drugs. This prompted the hypothesis of psychosis as a state of "endogenous" sensitization of the dopamine system although the exact basis of dopaminergic disturbances and the possible role of
Thiol-Michael coupling and ring-opening metathesis polymerization: facile access to functional exo-7-oxanorbornene dendron macromonomers.
Liu M, et al.
Polymer International, 63(7), 1174-1183 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service