716944
Gold nanowires
diam. × L 30 nm × 4,500 nm, dispersion (H2O), contains CTAB as stabilizer
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Empirical Formula (Hill Notation):
Au
Molecular Weight:
196.97
UNSPSC Code:
12352302
NACRES:
NA.23
Recommended Products
form
dispersion (H2O)
nanowires
Quality Level
contains
CTAB as stabilizer
concentration
≥50 μg/mL
diam. × L
30 nm × 4,500 nm
pH
3.0-5.0
density
1.00 g/mL at 25 °C
Mw/Mn
(<20% CV, monodispersity)
>95 (nanowires)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Physical properties
Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C
Legal Information
This product was produced under the methods claimed in U.S. Pat. Nos. 8,956,440 and 8,241,922
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service