Skip to Content
Merck
All Photos(5)

Documents

716952

Sigma-Aldrich

Gold nanowires

diam. × L 30 nm × 6,000 nm, dispersion (H2O)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
Molecular Weight:
196.97
UNSPSC Code:
12352302
NACRES:
NA.23

form

dispersion (H2O)
nanowires

contains

<3.5% CTAB as stabilizer

concentration

>50 μg/mL

diam. × L

30 nm × 6,000 nm

pH

3.0-5.0

density

1 g/mL at 25 °C

Mw/Mn

(<20% CV, monodispersity)
>95 (nanowires)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Physical properties

Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C

Legal Information

This product was produced under the methods claimed in U.S. Pat. Nos. 8,956,440 and 8,241,922

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service