Skip to Content
Merck
All Photos(2)

Documents

697745

Sigma-Aldrich

Cobalt

Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%

Synonym(s):

Cobalt element, Cobalt-59

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

material

carbon

Quality Level

Assay

≥99%

form

nanopowder

composition

Carbon content, <8 wt. %

magnetization

>150 emu/g, Mass saturation

resistivity

6.24 μΩ-cm, 20°C

surface area

>15 m2/g

particle size

<50 nm (TEM)

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Air- and thermally-stable magnetic carbon-coated cobalt nanoparticles (nanomagnets). Carbon shell consists of approximately three graphitic layers and can be chemically functionalized using e.g. diazotization reactions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun Ghee Tan et al.
Chemical communications (Cambridge, England), (36), 4297-4299 (2008-09-20)
To develop magnetic nanomaterials applicable to organic synthesis, the Suzuki cross-coupling method was adapted to attach a range of functional groups to carbon-coated core-shell materials via commercially-available substituted arylboronic acids.
Luechinger, N.A.; Booth, N.; Heness, G.; Bandyopadhyay, S.; Grass, R.N.; Stark W.J.
Advanced Materials, 20, 3044-3044 (2008)
Covalently functionalized cobalt nanoparticles as a platform for magnetic separations in organic synthesis.
Robert N Grass et al.
Angewandte Chemie (International ed. in English), 46(26), 4909-4912 (2007-05-23)
Alexander Schätz et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(27), 8262-8266 (2008-07-31)
TEMPO was grafted on graphene-coated nanobeads with a magnetic cobalt core by using a general applicable "click"-chemistry protocol. The new heterogeneous CoNP-TEMPO emerged as a highly active catalyst for the chemoselective oxidation of primary and secondary alcohols using bleach as
Jeannette Ø Penny et al.
Acta orthopaedica, 84(2), 130-137 (2013-04-20)
Wear particles from metal-on-metal arthroplasties are under suspicion for adverse effects both locally and systemically, and the DePuy ASR Hip Resurfacing System (RHA) has above-average failure rates. We compared lymphocyte counts in RHA and total hip arthroplasty (THA) and investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service