Skip to Content
Merck
All Photos(1)

Key Documents

746878

Sigma-Aldrich

Iron

nanopowder, 60-80 nm particle size, ≥99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99% trace metals basis

form

nanopowder

resistivity

9.71 μΩ-cm

particle size

60-80 nm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Application

Iron nanoparticles have been investigated for analytical applications including water purification of arsenic(1) and organic contaminants(2) and magnetic nanocomposites (3) as well as biomedical applications(4)

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Song; Hocheol;
Applied Catalysis. B, Environmental, 78(1-2), 53-60 (2008)
Sushil Raj Kanel et al.
Environmental science & technology, 39(5), 1291-1298 (2005-03-25)
Nanoscale zero-valent iron (NZVI) was synthesized and tested for the removal of As(III), which is a highly toxic, mobile, and predominant arsenic species in anoxic groundwater. We used SEM-EDX, AFM, and XRD to characterize particle size, surface morphology, and corrosion
Wilson;J. L.;
Journal of Applied Physics, 95(3), 1439-1443 (2004)
Kaori Kohara et al.
Chemical communications (Cambridge, England), 49(25), 2563-2565 (2013-02-21)
Carboxylated SiO2-coated α-Fe nanoparticles have been successfully prepared via CaH2-mediated reduction of SiO2-coated Fe3O4 nanoparticles followed by surface carboxylation. These α-Fe-based nanoparticles, which are characterized by ease of coating with additional functional groups, a large magnetization of 154 emu per

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service