Skip to Content
Merck
All Photos(2)

Key Documents

697575

Sigma-Aldrich

(Acetonitrile)[(2-biphenyl)di-tert-butylphosphine]gold(I) hexafluoroantimonate

Synonym(s):

JohnPhos Au(MeCN)SbF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30AuF6NPSb
CAS Number:
Molecular Weight:
772.17
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

198-203 °C (D)

SMILES string

[Au+].CC#N.F[Sb-](F)(F)(F)(F)F.CC(C)(C)P(c1ccccc1-c2ccccc2)C(C)(C)C

InChI

1S/C20H27P.C2H3N.Au.6FH.Sb/c1-19(2,3)21(20(4,5)6)18-15-11-10-14-17(18)16-12-8-7-9-13-16;1-2-3;;;;;;;;/h7-15H,1-6H3;1H3;;6*1H;/q;;+1;;;;;;;+5/p-6

InChI key

ADQOZROJMVWPRI-UHFFFAOYSA-H

General description

(Acetonitrile)[(2-biphenyl)di-tert-butylphosphine]gold(I) hexafluoroantimonate is a phosphine ligated gold(I) complex. It is used as a catalyst in a domino reaction to form C–C, C–N, and C–O bonds due to the ease with which it can activate C=C and C≡C bonds, thus allowing for unique rearrangements or reactions with various nucleophiles. Gold catalysis provides an excellent method to construct complex chemical architectures in a mild manner that would be difficult to achieve using other reaction paradigms.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service