Skip to Content
Merck
All Photos(4)

Documents

696277

Sigma-Aldrich

[(IPr)AuCl]

Umicore

Synonym(s):

Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I), 1,3-Bis(2,6-diisopropylphenyl-imidazol-2-ylidene)gold(I) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36AuClN2
CAS Number:
Molecular Weight:
621.01
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: gold
reagent type: catalyst

mp

298 °C

storage temp.

2-8°C

SMILES string

CC(C)c1cccc(C(C)C)c1N2C=CN(C2[Au]Cl)c3c(cccc3C(C)C)C(C)C

InChI

1S/C27H36N2.Au.ClH/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;;/h9-16,18-21H,1-8H3;;1H/q;+1;/p-1

InChI key

CNJQLSINQGKZAW-UHFFFAOYSA-M

Application

Gold Catalysts — 21st Century ′Gold Rush′

  • Catalyst use in the hydration of alkynes

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Marion et al.
Journal of the American Chemical Society, 131(2), 448-449 (2009-01-15)
A highly efficient [(NHC)Au(I)]-based (NHC = N-heterocyclic carbene) catalytic system for the hydration of an array of alkynes that operates under acid-free conditions and at very low catalyst loadings (typically 50-100 ppm and as low as 10 ppm) was developed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service